Learn More
Meso Scale Discovery UPLEX NHP Fractalkine ASY
SDP
Catalog No. 5000079890
Encompass
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human chemokine Fractalkine in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79890 Supplier Meso Scale Discovery Supplier No. K156VCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human chemokine Fractalkine in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL