Learn More
Meso Scale Discovery UPLEX NHP Fractalkine ASY384
SDP
Catalog No. 5000078395
Encompass
The Meso Scale Discovery U-PLEX NHP Fractalkine 384-Well Assay is a multiplex immunoassay kit for quantifying human chemokine Fractalkine in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs Requiring only 25 L per well the assay completes in about 4 hours The kit includes a 384-well plate with capture antibodies detection antibodies calibrators controls buffers and seals enabling efficient high-throughput Fractalkine screening
Catalog No. 50-000-78395 Supplier Meso Scale Discovery Supplier No. K256VCK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Fractalkine 384-Well Assay is a multiplex immunoassay kit for quantifying human chemokine Fractalkine in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs Requiring only 25 L per well the assay completes in about 4 hours The kit includes a 384-well plate with capture antibodies detection antibodies calibrators controls buffers and seals enabling efficient high-throughput Fractalkine screening