Learn More
Meso Scale Discovery UPLEX NHP Fractalkine ASY384
SDP
Catalog No. 5000079199
Encompass
The Meso Scale Discovery U-PLEX NHP Fractalkine 384-Well Assay Kit is a 384-well multiplex immunoassay kit for the quantitative measurement of human Fractalkine in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrators and assay diluent It allows simultaneous measurement of up to 16 samples per plate in under 4 hours
Catalog No. 50-000-79199 Supplier Meso Scale Discovery Supplier No. K256VCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Fractalkine 384-Well Assay Kit is a 384-well multiplex immunoassay kit for the quantitative measurement of human Fractalkine in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrators and assay diluent It allows simultaneous measurement of up to 16 samples per plate in under 4 hours