Learn More
Meso Scale Discovery UPLEX NHP GCSF ASY384well
SDP
Catalog No. 5000080220
Encompass
The U-PLEX NHP-G CSF 384-well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of neurofilament light chain (NfL) in human cerebrospinal fluid (CSF) It utilizes MSD s ultra-sensitive SULFO-TAG detection technology and is optimized for use on MSD s high-throughput 384-well MULTI-ARRAY and QUICKPLEX platforms The assay has a lower limit of quantification of 19 5 pg/mL and a working range up to 10 000 pg/mL It requires just 50 L of sample and can process up to 384 samples in 4 hours The assay provides sensitive reproducible quantitation of NfL a biomarker of neurodegeneration
Catalog No. 50-000-80220 Supplier Meso Scale Discovery Supplier No. K256VGK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP-G CSF 384-well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of neurofilament light chain (NfL) in human cerebrospinal fluid (CSF) It utilizes MSD s ultra-sensitive SULFO-TAG detection technology and is optimized for use on MSD s high-throughput 384-well MULTI-ARRAY and QUICKPLEX platforms The assay has a lower limit of quantification of 19 5 pg/mL and a working range up to 10 000 pg/mL It requires just 50 L of sample and can process up to 384 samples in 4 hours The assay provides sensitive reproducible quantitation of NfL a biomarker of neurodegeneration