Learn More
Meso Scale Discovery UPLEX NHP GMCSF ASY384well
SDP
Catalog No. 5000081238
Encompass
The U-PLEX NHP GM-CSF 384-well Assay Kit from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants The assay has a sensitivity of 0 07 pg/mL and a 7-point standard curve from 0 3-2000 pg/mL The kit includes a 384-well MULTI-ARRAY plate coated with GM-CSF capture antibody detection antibody read buffer wash buffer sample diluent and calibrators The assay takes approximately 3 5 hours to run
Catalog No. 50-000-81238 Supplier Meso Scale Discovery Supplier No. K256UMK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP GM-CSF 384-well Assay Kit from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants The assay has a sensitivity of 0 07 pg/mL and a 7-point standard curve from 0 3-2000 pg/mL The kit includes a 384-well MULTI-ARRAY plate coated with GM-CSF capture antibody detection antibody read buffer wash buffer sample diluent and calibrators The assay takes approximately 3 5 hours to run