Learn More
Meso Scale Discovery UPLEX NHP GRO ASY384well
SDP
Catalog No. 5000079201
Encompass
The Meso Scale Discovery Human GRO-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human GRO-alpha in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrator and sample diluent The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It uses MSD s SECTOR Imager for detection and is compatible with serum plasma tissue culture media and cell and tissue lysates offering a fast ultra-sensitive and robust assay for human GRO-alpha
Catalog No. 50-000-79201 Supplier Meso Scale Discovery Supplier No. K256UXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human GRO-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human GRO-alpha in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrator and sample diluent The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It uses MSD s SECTOR Imager for detection and is compatible with serum plasma tissue culture media and cell and tissue lysates offering a fast ultra-sensitive and robust assay for human GRO-alpha