Learn More
Meso Scale Discovery UPLEX NHP I309 ASY384well
SDP
Catalog No. 5000081745
Encompass
The Meso Scale Discovery U-PLEX NHP-I 309-384 Well Assay is a 384-well multiplex immunoassay panel that enables simultaneous quantification of up to 76 biomarkers including cytokines chemokines growth factors and immune modulators It utilizes electrochemiluminescent detection for high sensitivity and broad dynamic range The panel is optimized for analysis of non-human primate serum plasma and other biofluids It provides a streamlined workflow using as little as 12 5 L sample per well The U-PLEX platform delivers exceptional performance with accuracy precision and reproducibility for biomarker research
Catalog No. 50-000-81745 Supplier Meso Scale Discovery Supplier No. K256UYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP-I 309-384 Well Assay is a 384-well multiplex immunoassay panel that enables simultaneous quantification of up to 76 biomarkers including cytokines chemokines growth factors and immune modulators It utilizes electrochemiluminescent detection for high sensitivity and broad dynamic range The panel is optimized for analysis of non-human primate serum plasma and other biofluids It provides a streamlined workflow using as little as 12 5 L sample per well The U-PLEX platform delivers exceptional performance with accuracy precision and reproducibility for biomarker research