Learn More
Meso Scale Discovery UPLEX NHP IFN 2a ASY384well
SDP
Catalog No. 5000078396
Encompass
The Meso Scale Discovery U-PLEX NHP IFN alpha2a 384-well Assay Kit is designed to measure human interferon alpha 2a (IFN 2a) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of IFN 2a from 0 15-500 pg/mL The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and buffer for 384 tests This assay allows high-throughput screening and pharmacokinetic assessments
Catalog No. 50-000-78396 Supplier Meso Scale Discovery Supplier No. K256VHK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IFN alpha2a 384-well Assay Kit is designed to measure human interferon alpha 2a (IFN 2a) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of IFN 2a from 0 15-500 pg/mL The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and buffer for 384 tests This assay allows high-throughput screening and pharmacokinetic assessments