Learn More
Meso Scale Discovery UPLEX NHP IFN 2a ASY384well
SDP
Catalog No. 5000080221
Encompass
The Meso Scale Discovery U-PLEX NHP IFN alpha2a 384-well assay is a multiplex immunoassay kit for quantifying IFN alpha2a in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range The kit includes a 384-well plate with capture and detection antibodies buffers and calibrators It requires 50 L or less sample volume and can perform up to 40 tests The quantification range is 0 08-400 pg/mL for IFN alpha2a
Catalog No. 50-000-80221 Supplier Meso Scale Discovery Supplier No. K256VHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IFN alpha2a 384-well assay is a multiplex immunoassay kit for quantifying IFN alpha2a in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range The kit includes a 384-well plate with capture and detection antibodies buffers and calibrators It requires 50 L or less sample volume and can perform up to 40 tests The quantification range is 0 08-400 pg/mL for IFN alpha2a