Learn More
Meso Scale Discovery UPLEX NHP IFN ASY384well
SDP
Catalog No. 5000081222
Encompass
The Meso Scale Discovery U-PLEX NHP IFN gamma 384-well assay is an electrochemiluminescent immunoassay for measuring IFN gamma in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and broad range Detection limit is 0 04 pg/mL to 10 000 pg/mL It requires 12 5 L per well and takes 3 5 hours The kit includes a 384-well plate antibodies buffer diluent calibrators and control Suitable for non-human primate samples only
Catalog No. 50-000-81222 Supplier Meso Scale Discovery Supplier No. K256TTK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IFN gamma 384-well assay is an electrochemiluminescent immunoassay for measuring IFN gamma in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and broad range Detection limit is 0 04 pg/mL to 10 000 pg/mL It requires 12 5 L per well and takes 3 5 hours The kit includes a 384-well plate antibodies buffer diluent calibrators and control Suitable for non-human primate samples only