Learn More
Meso Scale Discovery UPLEX NHP IFN2a ASYQP
SDP
Catalog No. 5000080133
Encompass
The U-PLEX NHP IFN-alpha2a Assay (K156VHK) is a specialized immunoassay kit designed for quantitative measurement of interferon alpha-2a in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples making it valuable for preclinical studies and biomedical research involving interferon responses in primate models
Catalog No. 50-000-80133 Supplier Meso Scale Discovery Supplier No. K156VHK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IFN-alpha2a Assay (K156VHK) is a specialized immunoassay kit designed for quantitative measurement of interferon alpha-2a in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples making it valuable for preclinical studies and biomedical research involving interferon responses in primate models