Learn More
Meso Scale Discovery UPLEX NHP IL1 ASY384well
SDP
Catalog No. 5000081374
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1alpha 384-well Assay Kit is a multiplex immunoassay that measures human IL-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours The kit can quantify IL-1alpha concentrations between 0 05 and 1000 pg/mL
Catalog No. 50-000-81374 Supplier Meso Scale Discovery Supplier No. K256UNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1alpha 384-well Assay Kit is a multiplex immunoassay that measures human IL-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours The kit can quantify IL-1alpha concentrations between 0 05 and 1000 pg/mL