Learn More
Meso Scale Discovery UPLEX NHP IL10 ASY384well
SDP
Catalog No. 5000080201
Encompass
The Meso Scale Discovery U-PLEX NHP IL-10 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human interleukin 10 (IL-10) in cell culture supernatants serum plasma and other biological matrices This 384-well electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-10 Each kit provides sufficient reagents for analyzing 384 samples in a low-volume high-throughput microplate format
Catalog No. 50-000-80201 Supplier Meso Scale Discovery Supplier No. K256TZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-10 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human interleukin 10 (IL-10) in cell culture supernatants serum plasma and other biological matrices This 384-well electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-10 Each kit provides sufficient reagents for analyzing 384 samples in a low-volume high-throughput microplate format