Learn More
Meso Scale Discovery UPLEX NHP IL12/IL23p40 ASY384
SDP
Catalog No. 5000078393
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying human IL-12/IL-23p40 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume This multiplex assay measures IL-12 and IL-23 cytokines key in inflammatory and autoimmune responses suitable for biomarker research The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls
Catalog No. 50-000-78393 Supplier Meso Scale Discovery Supplier No. K256UQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying human IL-12/IL-23p40 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume This multiplex assay measures IL-12 and IL-23 cytokines key in inflammatory and autoimmune responses suitable for biomarker research The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls