Learn More
Meso Scale Discovery UPLEX NHP IL12/IL23p40 ASY384
SDP
Catalog No. 5000080202
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-12 and IL-23 in cell culture supernatants serum plasma and other biological samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It can detect IL-12 and IL-23 with lower limits of detection of 0 3 pg/mL and 0 2 pg/mL respectively The assay takes approximately 3 hours to complete
Catalog No. 50-000-80202 Supplier Meso Scale Discovery Supplier No. K256UQK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-12 and IL-23 in cell culture supernatants serum plasma and other biological samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It can detect IL-12 and IL-23 with lower limits of detection of 0 3 pg/mL and 0 2 pg/mL respectively The assay takes approximately 3 hours to complete