Learn More
Meso Scale Discovery UPLEX NHP IL12/IL23p40 ASYQP
SDP
Catalog No. 5000080709
Encompass
This is a MSD U-PLEX singleplex immunoassay designed to measure IL-12/IL-23 p40 protein levels in non-human primate samples The electrochemiluminescence-based platform provides high sensitivity and specificity for cytokine detection in research applications The assay utilizes MSD s proprietary MULTI-SPOT technology for reliable quantitative analysis of this inflammatory biomarker in primate biological samples
Catalog No. 50-000-80709 Supplier Meso Scale Discovery Supplier No. K156UQK22
This is a MSD U-PLEX singleplex immunoassay designed to measure IL-12/IL-23 p40 protein levels in non-human primate samples The electrochemiluminescence-based platform provides high sensitivity and specificity for cytokine detection in research applications The assay utilizes MSD s proprietary MULTI-SPOT technology for reliable quantitative analysis of this inflammatory biomarker in primate biological samples