Learn More
Meso Scale Discovery UPLEX NHP IL12p70 ASY 25 PL
SDP
Catalog No. 5000078541
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 37 pg/mL
Catalog No. 50-000-78541 Supplier Meso Scale Discovery Supplier No. K156UAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 37 pg/mL