Learn More
Meso Scale Discovery UPLEX NHP IL12p70 ASY384well
SDP
Catalog No. 5000080007
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12p70 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-12p70 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of IL-12p70 levels with a sample volume of 25 L The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrator
Catalog No. 50-000-80007 Supplier Meso Scale Discovery Supplier No. K256UAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-12p70 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-12p70 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of IL-12p70 levels with a sample volume of 25 L The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrator