Learn More
Meso Scale Discovery UPLEX NHP IL12p70 ASY384well
SDP
Catalog No. 5000081162
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12p70 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a detection range of 0 61-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls This kit allows high-throughput analysis of IL-12p70 levels with low sample volume requirements
Catalog No. 50-000-81162 Supplier Meso Scale Discovery Supplier No. K256UAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-12p70 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a detection range of 0 61-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls This kit allows high-throughput analysis of IL-12p70 levels with low sample volume requirements