Learn More
Meso Scale Discovery UPLEX NHP IL12p70 ASYQP
SDP
Catalog No. 5000080821
Encompass
The MSD U-PLEX IL-12p70 Singleplex Assay is a specialized immunoassay designed for measuring interleukin-12p70 levels in non-human primate samples This electrochemiluminescence-based platform offers high sensitivity and specificity for cytokine detection utilizing MSD s proprietary U-PLEX technology for reliable quantification The singleplex format allows focused analysis of IL-12p70 a key inflammatory cytokine involved in immune responses making it valuable for research applications in immunology and drug development studies using primate models
Catalog No. 50-000-80821 Supplier Meso Scale Discovery Supplier No. K156UAK24
Add to Cart
Add to Cart
Request a Quote
The MSD U-PLEX IL-12p70 Singleplex Assay is a specialized immunoassay designed for measuring interleukin-12p70 levels in non-human primate samples This electrochemiluminescence-based platform offers high sensitivity and specificity for cytokine detection utilizing MSD s proprietary U-PLEX technology for reliable quantification The singleplex format allows focused analysis of IL-12p70 a key inflammatory cytokine involved in immune responses making it valuable for research applications in immunology and drug development studies using primate models