Learn More
Meso Scale Discovery UPLEX NHP IL13 ASY384well
SDP
Catalog No. 5000081234
Encompass
The Meso Scale Discovery U-PLEX NHP IL-13 384-Well Assay Kit is a 384-well multiplex assay that measures human IL-13 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows for the accurate and reliable quantification of IL-13 levels in human samples The kit has a sensitivity of 0 04 pg/mL and requires a sample volume of 25 L
Catalog No. 50-000-81234 Supplier Meso Scale Discovery Supplier No. K256UBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-13 384-Well Assay Kit is a 384-well multiplex assay that measures human IL-13 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows for the accurate and reliable quantification of IL-13 levels in human samples The kit has a sensitivity of 0 04 pg/mL and requires a sample volume of 25 L