Learn More
Meso Scale Discovery UPLEX NHP IL15 ASY384well
SDP
Catalog No. 5000078394
Encompass
The Meso Scale Discovery U-PLEX NHP IL-15 384-Well Assay Kit is a 384-well multiplex assay that measures natural human IL-15 in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 384-well MULTI-ARRAY Plate Calibrator Blend Diluent 35 Read Buffer T (4X) and Sample Diluent It can detect IL-15 concentrations as low as 0 06 pg/mL with an upper limit of 10 000 pg/mL The total assay time is 3 5 hours This product enables efficient and sensitive analysis of natural human IL-15
Catalog No. 50-000-78394 Supplier Meso Scale Discovery Supplier No. K256URK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-15 384-Well Assay Kit is a 384-well multiplex assay that measures natural human IL-15 in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 384-well MULTI-ARRAY Plate Calibrator Blend Diluent 35 Read Buffer T (4X) and Sample Diluent It can detect IL-15 concentrations as low as 0 06 pg/mL with an upper limit of 10 000 pg/mL The total assay time is 3 5 hours This product enables efficient and sensitive analysis of natural human IL-15