Learn More
Meso Scale Discovery UPLEX NHP IL17A/F ASY 25 PL
SDP
Catalog No. 5000080063
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A/F Assay is a multiplex immunoassay kit for measuring IL-17A and IL-17F in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and broad dynamic range The assay requires 50 L sample volume and is compatible with MSD s SECTOR Imager instruments It includes a plate with capture and detection antibodies calibrators and controls for a standard curve The kit provides reagents for one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-80063 Supplier Meso Scale Discovery Supplier No. K156VYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17A/F Assay is a multiplex immunoassay kit for measuring IL-17A and IL-17F in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and broad dynamic range The assay requires 50 L sample volume and is compatible with MSD s SECTOR Imager instruments It includes a plate with capture and detection antibodies calibrators and controls for a standard curve The kit provides reagents for one 96-well plate testing 40 samples in duplicate