Learn More
Meso Scale Discovery UPLEX NHP IL17A/F ASY384well
SDP
Catalog No. 5000078423
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A/F 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17A and IL-17F in cell culture supernatants serum plasma and other biological matrices It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and minimal matrix interference in a 384-well plate format The assay requires a low sample volume of 50 L and has a short protocol time of approximately 3 5 hours Calibration curves controls and samples are measured in duplicate The lower limits of detection are 0 04 pg/mL for IL-17A and 0 17 pg/mL for IL-17F
Catalog No. 50-000-78423 Supplier Meso Scale Discovery Supplier No. K256VYK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17A/F 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17A and IL-17F in cell culture supernatants serum plasma and other biological matrices It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and minimal matrix interference in a 384-well plate format The assay requires a low sample volume of 50 L and has a short protocol time of approximately 3 5 hours Calibration curves controls and samples are measured in duplicate The lower limits of detection are 0 04 pg/mL for IL-17A and 0 17 pg/mL for IL-17F