Learn More
Meso Scale Discovery UPLEX NHP IL17B ASY384well
SDP
Catalog No. 5000078427
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17B 384-well assay kit measures IL-17B in human non-human primate rat and mouse samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a capture antibody-coated plate detection antibody with an electrochemiluminescent label calibrator and diluent It has a sensitivity of 0 3 pg/mL and an upper quantification limit of 10 000 pg/mL for human IL-17B
Catalog No. 50-000-78427 Supplier Meso Scale Discovery Supplier No. K256XNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17B 384-well assay kit measures IL-17B in human non-human primate rat and mouse samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a capture antibody-coated plate detection antibody with an electrochemiluminescent label calibrator and diluent It has a sensitivity of 0 3 pg/mL and an upper quantification limit of 10 000 pg/mL for human IL-17B