Learn More
Meso Scale Discovery UPLEX NHP IL17B ASYQP 25 PL
SDP
Catalog No. 5000078617
Encompass
The U-PLEX NHP IL-17B Assay (K156XNK) is a specialized immunoassay kit designed for quantitative measurement of Interleukin-17B in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying IL-17B levels in preclinical studies involving non-human primate models
Catalog No. 50-000-78617 Supplier Meso Scale Discovery Supplier No. K156XNK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IL-17B Assay (K156XNK) is a specialized immunoassay kit designed for quantitative measurement of Interleukin-17B in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying IL-17B levels in preclinical studies involving non-human primate models