Learn More
Meso Scale Discovery UPLEX NHP IL17C ASY384well
SDP
Catalog No. 5000079238
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17C 384-Well Assay is a multiplex immunoassay kit for measuring human IL-17C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection and features a 384-well plate with analyte-specific antibodies The detection range is 0 06 - 2500 pg/mL offering high sensitivity and a broad dynamic range with low sample volume Compatible with common sample types and automation the kit includes a 384-well plate detection antibodies read buffer sample diluent calibrators and control reagents
Catalog No. 50-000-79238 Supplier Meso Scale Discovery Supplier No. K256WJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17C 384-Well Assay is a multiplex immunoassay kit for measuring human IL-17C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection and features a 384-well plate with analyte-specific antibodies The detection range is 0 06 - 2500 pg/mL offering high sensitivity and a broad dynamic range with low sample volume Compatible with common sample types and automation the kit includes a 384-well plate detection antibodies read buffer sample diluent calibrators and control reagents