Learn More
Meso Scale Discovery UPLEX NHP IL17C ASYQP 25 PL
SDP
Catalog No. 5000080088
Encompass
The U-PLEX NHP IL-17C Assay (K156WJK) is a specialized immunoassay designed for measuring interleukin-17C levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of IL-17C a cytokine involved in inflammatory responses The assay is specifically validated for use with serum plasma and cell culture supernatants from non-human primate species making it valuable for preclinical research and drug development studies focusing on immune system function and inflammatory pathways
Catalog No. 50-000-80088 Supplier Meso Scale Discovery Supplier No. K156WJK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IL-17C Assay (K156WJK) is a specialized immunoassay designed for measuring interleukin-17C levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of IL-17C a cytokine involved in inflammatory responses The assay is specifically validated for use with serum plasma and cell culture supernatants from non-human primate species making it valuable for preclinical research and drug development studies focusing on immune system function and inflammatory pathways