Learn More
Meso Scale Discovery UPLEX NHP IL17D ASY384well
SDP
Catalog No. 5000079207
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17D 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 0 05-1000 pg/mL and includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate the standard curve and validate assay performance The kit provides reagents for testing up to 380 samples in duplicate
Catalog No. 50-000-79207 Supplier Meso Scale Discovery Supplier No. K256XOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17D 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 0 05-1000 pg/mL and includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate the standard curve and validate assay performance The kit provides reagents for testing up to 380 samples in duplicate