Learn More
Meso Scale Discovery UPLEX NHP IL17F ASY384well
SDP
Catalog No. 5000081754
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17F 384-Well Assay Kit is a 384-well multiplex immunoassay that measures natural human IL-17F in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 384-well high bind microplate It allows for the accurate quantitation of natural human IL-17F across a range of 15 6 - 1000 pg/mL
Catalog No. 50-000-81754 Supplier Meso Scale Discovery Supplier No. K256WAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17F 384-Well Assay Kit is a 384-well multiplex immunoassay that measures natural human IL-17F in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 384-well high bind microplate It allows for the accurate quantitation of natural human IL-17F across a range of 15 6 - 1000 pg/mL