Learn More
Meso Scale Discovery UPLEX NHP IL18 ASY384well
SDP
Catalog No. 5000079203
Encompass
The Meso Scale Discovery U-PLEX NHP IL-18 384-Well Assay is a 384-well multiplex assay kit that enables simultaneous quantification of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can accurately measure IL-18 concentrations from 0 61 pg/mL to 10 000 pg/mL
Catalog No. 50-000-79203 Supplier Meso Scale Discovery Supplier No. K256VJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-18 384-Well Assay is a 384-well multiplex assay kit that enables simultaneous quantification of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can accurately measure IL-18 concentrations from 0 61 pg/mL to 10 000 pg/mL