Learn More
Meso Scale Discovery UPLEX NHP IL1B ASY384well
SDP
Catalog No. 5000081214
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1beta 384-Well Assay is a multiplex immunoassay that measures human IL-1beta in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results in a 384-well plate format The assay requires a small sample volume and has a testing time of approximately 3 hours It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator and control solutions for quantitative analysis The assay is designed for research use only
Catalog No. 50-000-81214 Supplier Meso Scale Discovery Supplier No. K256TUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1beta 384-Well Assay is a multiplex immunoassay that measures human IL-1beta in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results in a 384-well plate format The assay requires a small sample volume and has a testing time of approximately 3 hours It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator and control solutions for quantitative analysis The assay is designed for research use only