Learn More
Meso Scale Discovery UPLEX NHP IL1RA ASYQP 25 PL
SDP
Catalog No. 5000080232
Encompass
The K156XPK-1 is an MSD electrochemiluminescence immunoassay kit for quantitative biomarker measurement in biological samples Using SULFO-TAG labels and carbon electrodes it ensures sensitive precise detection with minimal sample volume The kit includes pre-coated plates detection antibodies calibrators and reagents for a streamlined workflow Compatible with the MESO QuickPlex SQ 120 it supports biomarker analysis in drug discovery clinical research and diagnostics with enhanced reproducibility and reduced assay time
Catalog No. 50-000-80232 Supplier Meso Scale Discovery Supplier No. K156XPK24
Add to Cart
Add to Cart
Request a Quote
The K156XPK-1 is an MSD electrochemiluminescence immunoassay kit for quantitative biomarker measurement in biological samples Using SULFO-TAG labels and carbon electrodes it ensures sensitive precise detection with minimal sample volume The kit includes pre-coated plates detection antibodies calibrators and reagents for a streamlined workflow Compatible with the MESO QuickPlex SQ 120 it supports biomarker analysis in drug discovery clinical research and diagnostics with enhanced reproducibility and reduced assay time