Learn More
Meso Scale Discovery UPLEX NHP IL22 ASY384well
SDP
Catalog No. 5000080267
Encompass
The Meso Scale Discovery U-PLEX NHP IL-22 384-Well Assay Kit is a 384-well multiplex assay that measures human non-human primate and rodent IL-22 in serum plasma cell culture supernatants and other biological samples It utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and vials of lyophilized analyte standard It allows for the accurate quantitation of IL-22 levels with as little as 25 L of sample
Catalog No. 50-000-80267 Supplier Meso Scale Discovery Supplier No. K256WIK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-22 384-Well Assay Kit is a 384-well multiplex assay that measures human non-human primate and rodent IL-22 in serum plasma cell culture supernatants and other biological samples It utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and vials of lyophilized analyte standard It allows for the accurate quantitation of IL-22 levels with as little as 25 L of sample