Learn More
Meso Scale Discovery UPLEX NHP IL4 ASY384well
SDP
Catalog No. 5000081232
Encompass
The Meso Scale Discovery U-PLEX NHP IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring IL-4 in human non-human primate and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls Requiring only 25 L of sample it offers a dynamic range of 47-10 000 pg/mL aiding research into Th2-mediated immune responses
Catalog No. 50-000-81232 Supplier Meso Scale Discovery Supplier No. K256TWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring IL-4 in human non-human primate and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls Requiring only 25 L of sample it offers a dynamic range of 47-10 000 pg/mL aiding research into Th2-mediated immune responses