Learn More
Meso Scale Discovery UPLEX NHP IL6 ASY384well
SDP
Catalog No. 5000081226
Encompass
The Meso Scale Discovery U-PLEX NHP IL-6 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows high-throughput analysis of IL-6 levels with a small sample volume requirement of 25 L per well
Catalog No. 50-000-81226 Supplier Meso Scale Discovery Supplier No. K256TXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-6 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows high-throughput analysis of IL-6 levels with a small sample volume requirement of 25 L per well