Learn More
Meso Scale Discovery UPLEX NHP IL7 ASY384well
SDP
Catalog No. 5000081235
Encompass
The Meso Scale Discovery U-PLEX NHP IL-7 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay has a sensitivity of 0 11 pg/mL and a 7-point standard curve from 0 3 to 2000 pg/mL It uses MSD GOLD SULFO-TAG labels and carbon electrodes in microplates to provide sensitive reproducible results in just 3 5 hours The kit includes a plate detection antibody calibrator and assay diluent valid for 1 year from date of manufacture when stored as directed It is for research use only
Catalog No. 50-000-81235 Supplier Meso Scale Discovery Supplier No. K256UPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-7 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay has a sensitivity of 0 11 pg/mL and a 7-point standard curve from 0 3 to 2000 pg/mL It uses MSD GOLD SULFO-TAG labels and carbon electrodes in microplates to provide sensitive reproducible results in just 3 5 hours The kit includes a plate detection antibody calibrator and assay diluent valid for 1 year from date of manufacture when stored as directed It is for research use only