Learn More
Meso Scale Discovery UPLEX NHP IL8 ASY384well
SDP
Catalog No. 5000080199
Encompass
The Meso Scale Discovery U-PLEX NHP IL-8 384-Well Assay Kit is a 384-well multiplex assay that measures human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent It allows high-throughput screening and has a testing time of 3 5 hours The assay has a sensitivity of 0 06 pg/mL and requires a sample volume of 25 L
Catalog No. 50-000-80199 Supplier Meso Scale Discovery Supplier No. K256TYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-8 384-Well Assay Kit is a 384-well multiplex assay that measures human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent It allows high-throughput screening and has a testing time of 3 5 hours The assay has a sensitivity of 0 06 pg/mL and requires a sample volume of 25 L