Learn More
Meso Scale Discovery UPLEX NHP IL9 ASY384well
SDP
Catalog No. 5000078426
Encompass
The Meso Scale Discovery U-PLEX NHP IL-9 384-Well Assay Kit is a multiplex immunoassay that measures human IL-9 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format The assay requires 10 L of sample per well and the kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator Blend and Diluent It can detect IL-9 concentrations as low as 0 03 pg/mL with an upper limit of 10 000 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78426 Supplier Meso Scale Discovery Supplier No. K256XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-9 384-Well Assay Kit is a multiplex immunoassay that measures human IL-9 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format The assay requires 10 L of sample per well and the kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator Blend and Diluent It can detect IL-9 concentrations as low as 0 03 pg/mL with an upper limit of 10 000 pg/mL The assay takes approximately 3 5 hours to complete