Learn More
Meso Scale Discovery UPLEX NHP IP10 ASY384well
SDP
Catalog No. 5000079469
Encompass
The Meso Scale Discovery U-PLEX NHP IP-10 384-Well Assay Kit is a multiplex immunoassay that measures human IP-10 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate format with a testing range of 0 03-2500 pg/mL The assay requires 25 L of sample per well and the total assay time is approximately 3 5 hours This quantitative immunoassay allows high-throughput screening and aids in understanding IP-10 biology
Catalog No. 50-000-79469 Supplier Meso Scale Discovery Supplier No. K256UFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IP-10 384-Well Assay Kit is a multiplex immunoassay that measures human IP-10 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate format with a testing range of 0 03-2500 pg/mL The assay requires 25 L of sample per well and the total assay time is approximately 3 5 hours This quantitative immunoassay allows high-throughput screening and aids in understanding IP-10 biology