Learn More
Meso Scale Discovery UPLEX NHP MCSF ASY384well
SDP
Catalog No. 5000081771
Encompass
The U-PLEX NHP CSF 384-well Assay from Meso Scale Diagnostics measures 38 biomarkers in human CSF using electrochemiluminescence on the 384-well MESO QuickPlex SQ 120 It assesses neurodegenerative neuroinflammatory and neurologic disease markers like A peptides tau -synuclein NFL and GFAP Requiring 12 5 L per well it tests up to 40 samples in duplicate aiding in diagnosing Alzheimer s Parkinson s and other disorders
Catalog No. 50-000-81771 Supplier Meso Scale Discovery Supplier No. K256XRK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP CSF 384-well Assay from Meso Scale Diagnostics measures 38 biomarkers in human CSF using electrochemiluminescence on the 384-well MESO QuickPlex SQ 120 It assesses neurodegenerative neuroinflammatory and neurologic disease markers like A peptides tau -synuclein NFL and GFAP Requiring 12 5 L per well it tests up to 40 samples in duplicate aiding in diagnosing Alzheimer s Parkinson s and other disorders