Learn More
Meso Scale Discovery UPLEX NHP MDC ASY384well
SDP
Catalog No. 5000081229
Encompass
Meso Scale Discovery offers the V-PLEX NHP Cytokine Panel 1 (non-human primate) Kit which is a multiplex immunoassay that measures proinflammatory cytokines and chemokines in non-human primate samples such as serum plasma and culture supernatants This assay allows users to quantify biomarkers related to inflammation and immune response The panel includes IFN-gamma IL-1beta IL-6 IL-8 IL-10 IL-12/23p40 MCP-1 and TNF-alpha The electrochemiluminescence technology provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This product can help researchers study immune function and inflammation in non-human primate models of human disease
Catalog No. 50-000-81229 Supplier Meso Scale Discovery Supplier No. K256UIK4
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers the V-PLEX NHP Cytokine Panel 1 (non-human primate) Kit which is a multiplex immunoassay that measures proinflammatory cytokines and chemokines in non-human primate samples such as serum plasma and culture supernatants This assay allows users to quantify biomarkers related to inflammation and immune response The panel includes IFN-gamma IL-1beta IL-6 IL-8 IL-10 IL-12/23p40 MCP-1 and TNF-alpha The electrochemiluminescence technology provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This product can help researchers study immune function and inflammation in non-human primate models of human disease