Learn More
Meso Scale Discovery UPLEX NHP MIF ASY384well
SDP
Catalog No. 5000080268
Encompass
The U-PLEX NHP MIF 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that enables simultaneous detection of up to 10 biomarkers per well from a small sample volume It utilizes electrochemiluminescence for high sensitivity and broad dynamic range This assay measures human MIF in cell culture supernatants serum plasma and other biological matrices with a lower limit of detection of 45 5 pg/mL and an upper limit of quantification of 50 000 pg/mL The kit includes a 384-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls Recommended sample volume is 25 L
Catalog No. 50-000-80268 Supplier Meso Scale Discovery Supplier No. K256XJK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MIF 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that enables simultaneous detection of up to 10 biomarkers per well from a small sample volume It utilizes electrochemiluminescence for high sensitivity and broad dynamic range This assay measures human MIF in cell culture supernatants serum plasma and other biological matrices with a lower limit of detection of 45 5 pg/mL and an upper limit of quantification of 50 000 pg/mL The kit includes a 384-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls Recommended sample volume is 25 L