Learn More
Meso Scale Discovery UPLEX NHP MIP1B ASY384well
SDP
Catalog No. 5000081373
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1beta 384-well assay kit is a multiplex immunoassay that enables simultaneous quantification of the human chemokine MIP-1beta in serum plasma and cell culture supernatants The kit contains a 384-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve The electrochemiluminescent readout provides high sensitivity and a wide dynamic range This assay allows high-throughput analysis of MIP-1beta levels with low sample volume requirements
Catalog No. 50-000-81373 Supplier Meso Scale Discovery Supplier No. K256UKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-1beta 384-well assay kit is a multiplex immunoassay that enables simultaneous quantification of the human chemokine MIP-1beta in serum plasma and cell culture supernatants The kit contains a 384-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve The electrochemiluminescent readout provides high sensitivity and a wide dynamic range This assay allows high-throughput analysis of MIP-1beta levels with low sample volume requirements