Learn More
Meso Scale Discovery UPLEX NHP MIP3B ASY384well
SDP
Catalog No. 5000079197
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 384-well Assay Kit is a multiplex immunoassay kit for the quantitative measurement of human MIP-3 in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity and wide dynamic range This product requires the MSD reader for analysis It includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-79197 Supplier Meso Scale Discovery Supplier No. K256VAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-3 384-well Assay Kit is a multiplex immunoassay kit for the quantitative measurement of human MIP-3 in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity and wide dynamic range This product requires the MSD reader for analysis It includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance