Learn More
Meso Scale Discovery UPLEX NHP MIP5 ASY384well
SDP
Catalog No. 5000080272
Encompass
The U-PLEX NHP MIP 5-Plex 384-well Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay panel for measuring 5 biomarkers (A 40 A 42 -synuclein p-tau t-tau) in human CSF and plasma using electrochemiluminescent detection It features Meso Scale s Multi-Array technology with carbon ink electrodes The kit includes a plate with capture antibodies detection antibodies calibrators and controls for the 5-plex assay offering sensitive reproducible results from 25 L samples in a high-throughput format
Catalog No. 50-000-80272 Supplier Meso Scale Discovery Supplier No. K256XSK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MIP 5-Plex 384-well Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay panel for measuring 5 biomarkers (A 40 A 42 -synuclein p-tau t-tau) in human CSF and plasma using electrochemiluminescent detection It features Meso Scale s Multi-Array technology with carbon ink electrodes The kit includes a plate with capture antibodies detection antibodies calibrators and controls for the 5-plex assay offering sensitive reproducible results from 25 L samples in a high-throughput format