Learn More
Meso Scale Discovery UPLEX NHP SDF1 ASY384well
SDP
Catalog No. 5000079198
Encompass
SDF-1 is a chemokine produced by stromal cells that binds to CXCR4 receptors on hematopoietic stem cells lymphocytes and cancer cells It plays a role in stem cell homing lymphocyte trafficking angiogenesis and cancer metastasis SDF-1 can be detected in biological samples using ELISA kits from Meso Scale Discovery which utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The kits include standards controls plates detection antibodies read buffer and wash buffer for quantitative measurement of human mouse and rat SDF-1
Catalog No. 50-000-79198 Supplier Meso Scale Discovery Supplier No. K256VBK4
Add to Cart
Add to Cart
Request a Quote
SDF-1 is a chemokine produced by stromal cells that binds to CXCR4 receptors on hematopoietic stem cells lymphocytes and cancer cells It plays a role in stem cell homing lymphocyte trafficking angiogenesis and cancer metastasis SDF-1 can be detected in biological samples using ELISA kits from Meso Scale Discovery which utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The kits include standards controls plates detection antibodies read buffer and wash buffer for quantitative measurement of human mouse and rat SDF-1