Learn More
Meso Scale Discovery UPLEX NHP SDF1 ASYQP 25 PL
SDP
Catalog No. 5000080127
Encompass
The U-PLEX NHP SDF-1alpha Assay (K156VBK) is a specialized immunoassay kit designed for quantitative measurement of SDF-1alpha (stromal cell-derived factor-1 alpha) in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples providing researchers with a reliable tool for studying chemokine biology and inflammatory responses in preclinical studies
Catalog No. 50-000-80127 Supplier Meso Scale Discovery Supplier No. K156VBK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP SDF-1alpha Assay (K156VBK) is a specialized immunoassay kit designed for quantitative measurement of SDF-1alpha (stromal cell-derived factor-1 alpha) in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples providing researchers with a reliable tool for studying chemokine biology and inflammatory responses in preclinical studies