Learn More
Meso Scale Discovery UPLEX NHP TARC ASY384well
SDP
Catalog No. 5000079471
Encompass
The U-PLEX NHP TARC 384-well Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal matrix interference The kit includes a 384-well plate detection antibody calibrators and necessary reagents Results are analyzed on MSD s platforms for precise TARC quantitation For research use only
Catalog No. 50-000-79471 Supplier Meso Scale Discovery Supplier No. K256ULK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP TARC 384-well Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal matrix interference The kit includes a 384-well plate detection antibody calibrators and necessary reagents Results are analyzed on MSD s platforms for precise TARC quantitation For research use only