Learn More
Meso Scale Discovery UPLEX NHP TGFB1 ASYQP 25 PL
SDP
Catalog No. 5000081078
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta1 Assay is an electrochemiluminescent immunoassay for measuring active and total TGF-beta1 in non-human primate serum plasma and cell culture supernatants It uses U-PLEX technology to measure TGF-beta1 and other biomarkers from 25 L samples The detection range is 1 3 to 10 000 pg/mL The kit includes a 96-well plate antibodies calibrators and reagents with a 3 5-hour turnaround It offers sensitive reproducible quantitation of TGF-beta1 in NHP samples
Catalog No. 50-000-81078 Supplier Meso Scale Discovery Supplier No. K156XWK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta1 Assay is an electrochemiluminescent immunoassay for measuring active and total TGF-beta1 in non-human primate serum plasma and cell culture supernatants It uses U-PLEX technology to measure TGF-beta1 and other biomarkers from 25 L samples The detection range is 1 3 to 10 000 pg/mL The kit includes a 96-well plate antibodies calibrators and reagents with a 3 5-hour turnaround It offers sensitive reproducible quantitation of TGF-beta1 in NHP samples